4-(2,3-Dihydro-1H-inden-5-yl)pyrimidin-2-amine, tech
Catalog # APOSOR33038-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.262 g/mol
Storage Temperature:Ambient
CAS Number:866050-31-9
Formula:C₁₃H₁₃N₃
MDL Number:MFCD03787582