[5-(2-Chlorophenyl)-1,2-oxazol-3-yl]methanol, tech
Catalog # APOSOR33004-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:209.629 g/mol
Storage Temperature:Ambient
CAS Number:343374-44-7
Formula:C₁₀H₈ClNO₂
MDL Number:MFCD01444102