6-[(Pyridin-2-yl)methyl]pyridazin-3-ol, tech
Catalog # APOSOR32951-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.198 g/mol
Storage Temperature:Ambient
CAS Number:338779-41-2
Formula:C₁₀H₉N₃O
MDL Number:MFCD00140707