1-Acetyl-2,2,6,6-tetramethylpiperidin-4-one, tech
Catalog # APOSOR32908-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.274 g/mol
Storage Temperature:Ambient
CAS Number:52326-65-5
Formula:C₁₁H₁₉NO₂
MDL Number:MFCD02571209