1-(2-Phenylethoxy)-1H-imidazole-2-carbaldehyde, tech
Catalog # APOSOR32894-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.236 g/mol
Storage Temperature:Ambient
CAS Number:478050-31-6
Formula:C₁₂H₁₂N₂O₂
MDL Number:MFCD01871524