(2E)-3-[(3,5-Dichlorophenyl)amino]prop-2-enenitrile, tech
Catalog # APOSOR32863-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.063 g/mol
Storage Temperature:Ambient
CAS Number:338959-87-8
Formula:C₉H₆Cl₂N₂
MDL Number:MFCD00663724