Order Entry
Netherlands
ContactUsLinkComponent
(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal, tech
(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal, tech
Catalog # APOSOR32858-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal, tech
Catalog # APOSOR32858-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.117 g/mol
Storage Temperature:Ambient
CAS Number:866038-55-3
Formula:C₁₁H₁₁Cl₂NO
MDL Number:MFCD04125588