Order Entry
Netherlands
ContactUsLinkComponent
4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide, tech
4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide, tech
Catalogus # APOSOR32775-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide, tech
Catalogus # APOSOR32775-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.213 g/mol
Storage Temperature:Ambient
CAS Number:322414-12-0
Formula:C₁₀H₁₀N₄O
MDL Number:MFCD00448052