Methyl 4-[(6-chloropyridin-3-yl)methoxy]benzoate, tech
Catalog # APOSOR32699-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.703 g/mol
Storage Temperature:Ambient
CAS Number:865660-08-8
Formula:C₁₄H₁₂ClNO₃
MDL Number:MFCD04125299