Order Entry
Netherlands
ContactUsLinkComponent
 
N-(3,4-Dimethoxyphenyl)-2-methylpiperidine-1-carboxamide, tech
Catalog # APOSOR32694-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(3,4-Dimethoxyphenyl)-2-methylpiperidine-1-carboxamide, tech
Catalog # APOSOR32694-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.347 g/mol
Storage Temperature:Ambient
CAS Number:865659-53-6
Formula:C₁₅H₂₂N₂O₃
MDL Number:MFCD04125232