Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(4-Chlorophenyl)-2-oxopropanamide
N-(4-Chlorophenyl)-2-oxopropanamide, tech
N-(4-Chlorophenyl)-2-oxopropanamide, tech
APOSOR32623-1G
:
Apollo Scientific
:
N-(4-Chlorophenyl)-2-oxopropanamide, tech
APOSOR32623-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
197.618 g/mol
:
Ambient
:
52181-18-7
:
C₉H₈ClNO₂
:
MFCD02571800