Order Entry
Netherlands
ContactUsLinkComponent
Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Catalog # APOSOR32615-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Catalog # APOSOR32615-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.11 g/mol
Storage Temperature:Ambient
CAS Number:129109-19-9
Formula:C₁₂H₈Cl₂N₂O₃
MDL Number:MFCD00141184