Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 4-[(1-oxo-2,3-dihydro-1H-isoindol-2-yl)methyl]benzoate, tech
Ethyl 4-[(1-oxo-2,3-dihydro-1H-isoindol-2-yl)methyl]benzoate, tech
Catalog # APOSOR32595-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-[(1-oxo-2,3-dihydro-1H-isoindol-2-yl)methyl]benzoate, tech
Catalog # APOSOR32595-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :295.332 g/mol
Storage Temperature:Ambient
CAS Number:861211-67-8
Formula:C₁₈H₁₇NO₃
MDL Number:MFCD06496169