Order Entry
Netherlands
ContactUsLinkComponent
(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine, tech
(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine, tech
Catalog # APOSOR32565-1G
Supplier:  Apollo Scientific
CAS Number:  
(1Z,2E)-3-Phenyl-N-(4H-1,2,4-triazol-4-yl)prop-2-en-1-imine, tech
Catalog # APOSOR32565-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.224 g/mol
Storage Temperature:Ambient
CAS Number:35554-48-4
Formula:C₁₁H₁₀N₄
MDL Number:MFCD01326436