Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Chlorophenyl)-3-hydroxy-3-methylurea, tech
1-(4-Chlorophenyl)-3-hydroxy-3-methylurea, tech
Catalog # APOSOR32445-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Chlorophenyl)-3-hydroxy-3-methylurea, tech
Catalog # APOSOR32445-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.622 g/mol
Storage Temperature:Ambient
CAS Number:26816-99-9
Formula:C₈H₉ClN₂O₂
MDL Number:MFCD01814620