Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate, tech
Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate, tech
Catalog # APOSOR32438-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-[(3E)-2-oxo-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-3-ylidene]acetate, tech
Catalog # APOSOR32438-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.285 g/mol
Storage Temperature:Ambient
CAS Number:1308732-57-1
Formula:C₁₅H₁₅NO₃
MDL Number:MFCD01568302