Order Entry
Netherlands
ContactUsLinkComponent
 
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(piperidin-1-yl)propan-2-ol, tech
Catalog # APOSOR32432-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(piperidin-1-yl)propan-2-ol, tech
Catalog # APOSOR32432-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :288.385 g/mol
Storage Temperature:Ambient
CAS Number:338392-20-4
Formula:C₁₇H₂₄N₂O₂
MDL Number:MFCD01315615