1-(3-Chlorophenyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one, tech
Catalog # APOSOR32391-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.664 g/mol
Storage Temperature:Ambient
CAS Number:320419-78-1
Formula:C₁₂H₈ClN₃O
MDL Number:MFCD00140663