Order Entry
Netherlands
ContactUsLinkComponent
N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine 97%
N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine 97%
Catalog # APOSOR32369-1G
Supplier:  Apollo Scientific
CAS Number:  
N4,N4-Dimethyl-5-phenylpyrimidine-2,4-diamine 97%
Catalog # APOSOR32369-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.266 g/mol
Storage Temperature:Ambient
CAS Number:320424-61-1
Formula:C₁₂H₁₄N₄
MDL Number:MFCD00127225