Order Entry
Netherlands
ContactUsLinkComponent
2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Catalog # APOSOR32349-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Catalog # APOSOR32349-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.25 g/mol
Storage Temperature:Ambient
CAS Number:860785-01-9
Formula:C₁₂H₁₂N₄O
MDL Number:MFCD03787670