Order Entry
Netherlands
ContactUsLinkComponent
2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech
2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech
Catalog # APOSOR32333-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech
Catalog # APOSOR32333-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.322 g/mol
Storage Temperature:Ambient
CAS Number:477864-32-7
Formula:C₁₇H₁₇NO₂
MDL Number:MFCD02082957