(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one, tech
Catalog # APOSOR32292-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.722 g/mol
Storage Temperature:Ambient
CAS Number:303996-62-5
Formula:C₁₃H₁₃ClN₄O
MDL Number:MFCD00215245