Order Entry
Netherlands
ContactUsLinkComponent
2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
Catalog # APOSOR32258-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
Catalog # APOSOR32258-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.199 g/mol
Storage Temperature:Ambient
CAS Number:161176-99-4
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD03617589