(3Z)-1-Methyl-3-(2-phenylhydrazin-1-ylidene)-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSOR32209-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.283 g/mol
Storage Temperature:Ambient
CAS Number:15096-16-9
Formula:C₁₅H₁₃N₃O
MDL Number:MFCD00194472