5-Acetyl-6-methyl-2-(piperidin-1-yl)pyridine-3-carbonitrile, tech
Catalog # APOSOR32204-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.304 g/mol
Storage Temperature:Ambient
CAS Number:121348-11-6
Formula:C₁₄H₁₇N₃O
MDL Number:MFCD00793588