Methyl 1-(pyrimidin-2-yl)piperidine-4-carboxylate, tech
Catalog # APOSOR32195-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.256 g/mol
Storage Temperature:Ambient
CAS Number:303144-42-5
Formula:C₁₁H₁₅N₃O₂
MDL Number:MFCD00664472