4-(3-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
Catalog # APOSOR32184-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.714 g/mol
Storage Temperature:Ambient
CAS Number:64123-84-8
Formula:C₁₅H₁₁ClN₂O
MDL Number:MFCD00232641