Ethyl (R)-4-(piperidin-2-yl)benzoate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2307737-09-1
:
Formula:C₁₄H₁₉NO₂·ClH
MDL Number:MFCD30738189
:269.767 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together