Order Entry
Netherlands
ContactUsLinkComponent
3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol, tech
3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol, tech
Catalog # APOSOR32140-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol, tech
Catalog # APOSOR32140-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.365 g/mol
Storage Temperature:Ambient
CAS Number:860644-63-9
Formula:C₁₈H₂₁NO₂
MDL Number:MFCD04124253