N-(4-(Piperidin-4-yl)phenyl)acetamide hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2064217-79-2
:
Formula:C₁₃H₁₈N₂O·ClH
MDL Number:MFCD29075576
:254.756 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together