Order Entry
Netherlands
ContactUsLinkComponent
2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol, tech
2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol, tech
Catalog # APOSOR32137-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(2,3,4,9-Tetrahydro-1H-carbazol-1-yl)amino]ethan-1-ol, tech
Catalog # APOSOR32137-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.306 g/mol
Storage Temperature:Ambient
CAS Number:174152-39-7
Formula:C₁₄H₁₈N₂O
MDL Number:MFCD04124240