Order Entry
Netherlands
ContactUsLinkComponent
(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSOR32136-1G
Supplier:  Apollo Scientific
CAS Number:  
(3Z)-3-[(2-Aminophenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
Catalog # APOSOR32136-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.269 g/mol
Storage Temperature:Ambient
CAS Number:17990-71-5
Formula:C₁₅H₁₂N₂O
MDL Number:MFCD04124213