Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate 97%
Corrosive
Irritant
tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321348-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321348-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.331 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:259180-77-3
Formula:C₁₂H₂₄N₂O₂
MDL Number:MFCD04038458