
Ethyl (R)-3-(piperidin-2-yl)benzoate hydrochloride 97%
Catalog # APOSOR321270-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:269.767 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:2244064-28-4
Formula:C₁₄H₁₉NO₂·ClH
MDL Number:MFCD30099273