Order Entry
Netherlands
ContactUsLinkComponent
N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 90%
Irritant
N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 90%
Catalog # APOSOR321253-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N,N-Dimethyl-1-(2-(piperidin-4-yl)phenyl)methanamine dihydrochloride 90%
Catalog # APOSOR321253-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :291.26 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:2244087-45-2
Formula:C₁₄H₂₂N₂·2ClH
MDL Number:MFCD30099254