
N,N-Dimethyl-1-(4-(piperidin-4-yl)phenyl)methanamine dihydrochloride 97%
Catalog # APOSOR321252-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:291.26 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:2244087-73-6
Formula:C₁₄H₂₂N₂·2ClH
MDL Number:MFCD30099253