Order Entry
Netherlands
ContactUsLinkComponent
Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride 97%
Irritant
Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride 97%
Catalog # APOSOR321209-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl (R)-2-(piperidin-2-yl)acetate hydrochloride 97%
Catalog # APOSOR321209-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :207.698 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1000291-88-2
Formula:C₉H₁₇NO₂·ClH
MDL Number:MFCD13184088