Order Entry
Netherlands
ContactUsLinkComponent
4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one 95%
4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32003-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(Dimethylamino)methylidene]-3-(methoxymethyl)-4,5-dihydro-1H-pyrazol-5-one 95%
Catalog # APOSOR32003-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.208 g/mol
Storage Temperature:Ambient
CAS Number:240799-57-9
Formula:C₈H₁₃N₃O₂
MDL Number:MFCD00138266