
1-Boc-3-hydroxy-3-(aminomethyl)azetidine
Catalog # APOSOR317175-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.251 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1008526-71-3
Formula:C₉H₁₈N₂O₃
MDL Number:MFCD17015986