
3-(Bromomethyl)-7-chlorobenzo[b]thiophene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17512-61-7
:
Formula:C₉H₆BrClS
MDL Number:MFCD07787461
:261.566 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together