
(1R,4R)-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
:134003-84-2
:
:C₁₀H₁₈N₂O₂
:MFCD01321293
:198.262 g/mol
:75…77 °C
:GHS07
:Refrigerator
Frequently Bought Together
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Phosphorus standard dissolved in oil, 5,000 µg/g P in mineral oil for wear metal analysis, Specpure®
: Thermo Fisher Scientific