Order Entry
Netherlands
ContactUsLinkComponent
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%
Catalog # APOSOR315434-1G
Supplier:  Apollo Scientific
CAS Number:  
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%
Catalog # APOSOR315434-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.776 g/mol
Storage Temperature:Ambient
CAS Number:929302-18-1
Formula:C₁₂H₂₂N₂O₂·ClH
MDL Number:MFCD09835396