(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%
Catalog # APOSOR315137-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:250 mg
- Seq:2590
Specifications
About this item
:
:165.618 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:359849-58-4
Formula:C₆H₁₁NO₂·ClH
MDL Number:MFCD09749902