1-Methyl-3-phenyl-2,5,6,7-tetrahydro-1H-1,4-diazepin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:87517-47-3
:
Formula:C₁₂H₁₄N₂O
MDL Number:MFCD28122586
:202.252 g/mol
Storage Temperature:Ambient
Frequently Bought Together