Order Entry
Netherlands
ContactUsLinkComponent
3-Anilino-5,5-dimethyl-2-cyclohexen-1-one
3-Anilino-5,5-dimethyl-2-cyclohexen-1-one
Catalog # APOSOR311067-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Anilino-5,5-dimethyl-2-cyclohexen-1-one
Catalog # APOSOR311067-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.291 g/mol
Storage Temperature:Ambient
CAS Number:18940-21-1
Formula:C₁₄H₁₇NO
MDL Number:MFCD00128479