Order Entry
Netherlands
Orders LinkContactUsLinkComponent
[3-(Methylamino)phenyl]methanol

[3-(Methylamino)phenyl]methanol

Supplier:   Apollo Scientific
[3-(Methylamino)phenyl]methanol
Supplier:  Apollo Scientific
Certificates
[3-(Methylamino)phenyl]methanol

2 Options Available Below

Product Details & Documents
  • CAS Number:121562-78-5
  • Formula:C₈H₁₁NO
  • MDL Number:MFCD16658585
  • Molecular Weight:137.179 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 780,00 €
    780,00 €
    1 * 500 mg
       780,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 230,00 €
    1 230,00 €
    1 * 1 g
       1 230,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.