Order Entry
Netherlands
ContactUsLinkComponent
3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid
3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid
Catalog # APOSOR310782-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid
Catalog # APOSOR310782-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.269 g/mol
Storage Temperature:Ambient
CAS Number:1242874-27-6
Formula:C₁₂H₈N₂O₂S
MDL Number:MFCD15732197