Ethyl 5-(3,5-dichlorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Catalog # APOSOR310536-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.153 g/mol
Storage Temperature:Ambient
CAS Number:1565827-84-0
Formula:C₁₃H₁₂Cl₂N₂O₂
MDL Number:MFCD27957340