Methyl 1-[2-(2H-1,3-benzodioxol-5-yl)ethyl]-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Catalogus # APOSOR310504-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:318.281 g/mol
Melting Pt:226…228 °C
Storage Temperature:Ambient
CAS Number:1565827-86-2
Formula:C₁₅H₁₄N₂O₆
MDL Number:MFCD27957327