Methyl 2-(2-cyano-3-ethoxy-3-oxopropyl)benzoate
Catalog # APOSOR310407-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:261.273 g/mol
Storage Temperature:Ambient
CAS Number:153802-16-5
Formula:C₁₄H₁₅NO₄
MDL Number:MFCD06211147